Learn More
Medchemexpress LLC Mal-amido-PEG12-NHS ester | 2101722-60-3 | 98.52% | C38H63N3O19 | 50 MG
SDP
Catalog No. 5000353172
Encompass_Preferred
Mal-amido-PEG12-NHS ester is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. These PROTACs are composed of two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to a target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of specific target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Achieves selective degradation of target proteins
Catalog No. 50-003-53172 Supplier Medchemexpress LLC Supplier No. HY14097750MG
Mal-amido-PEG12-NHS ester is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. These PROTACs are composed of two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to a target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of specific target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Achieves selective degradation of target proteins