Learn More
Medchemexpress LLC Mal-amido-PEG12-NHS ester | 2101722-60-3 | MFCD11041148 | 98.5% | C38H63N3O19 | 100 MG
SDP
Catalog No. 5000354896
Encompass_Preferred
Mal-amido-PEG12-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs.
  • Protacs consist of two distinct ligands connected by a linker.
  • One ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest.
  • Protacs leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
Catalog No. 50-003-54896 Supplier Medchemexpress LLC Supplier No. HY140977100MG
Mal-amido-PEG12-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs.
  • Protacs consist of two distinct ligands connected by a linker.
  • One ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest.
  • Protacs leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.