Learn More
Medchemexpress LLC Mal-amido-PEG2-C2-acid | 756525-98-1 | 99.8% | 328.32 | 1 G
SDP
Catalog No. 5000405211
Encompass_Preferred
Mal-amido-PEG2-C2-acid is a PROTAC linker that is PEG-based and can be utilized in the synthesis of PROTACs. PROTACs themselves consist of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a protein of interest. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. This product is intended for research use only and is not sold to patients.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
Catalog No. 50-004-05211 Supplier Medchemexpress LLC Supplier No. HY421511G
Mal-amido-PEG2-C2-acid is a PROTAC linker that is PEG-based and can be utilized in the synthesis of PROTACs. PROTACs themselves consist of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a protein of interest. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. This product is intended for research use only and is not sold to patients.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs