Learn More
Medchemexpress LLC Mal-amido-PEG2-NHS ester | 955094-26-5 | MFCD11041137 | 99.3% | 425.39 g/mol | C18H23N3O9 | 50 MG
SDP
Catalog No. 5000204170
Encompass_Preferred
Mal-amido-PEG2-NHS ester is a noncleavable ADC linker containing a maleimide reactive group and an N-hydroxysuccinimide (NHS) ester. The NHS ester labels primary amines (-NH2) to form stable amide bonds, while the maleimide enables thiol conjugation for bioconjugation and linker chemistry applications.
  • Noncleavable ADC linker with maleimide and NHS ester.
  • Reacts with primary amines to form stable amide bonds.
  • Enables thiol conjugation via the maleimide functionality.
  • High purity suitable for research-grade bioconjugation.
  • Supplied as a solid for easy handling and storage.
Catalog No. 50-002-04170 Supplier Medchemexpress LLC Supplier No. HYW04028950MG
Mal-amido-PEG2-NHS ester is a noncleavable ADC linker containing a maleimide reactive group and an N-hydroxysuccinimide (NHS) ester. The NHS ester labels primary amines (-NH2) to form stable amide bonds, while the maleimide enables thiol conjugation for bioconjugation and linker chemistry applications.
  • Noncleavable ADC linker with maleimide and NHS ester.
  • Reacts with primary amines to form stable amide bonds.
  • Enables thiol conjugation via the maleimide functionality.
  • High purity suitable for research-grade bioconjugation.
  • Supplied as a solid for easy handling and storage.