Learn More
Medchemexpress LLC Mal-amido-PEG2-Val-Cit-PAB-PNP | 2112738-13-1 | 98.4% | 854.86 g/mol | C39H50N8O14 | 50 MG
SDP
Catalog No. 5000203813
Encompass_Preferred
Mal-amido-PEG2-Val-Cit-PAB-PNP is a cleavable two-unit PEG linker designed for use in the synthesis of antibody-drug conjugates (ADCs). It contains a val-cit dipeptide cleavable moiety, a p-aminobenzyl (PAB) self-immolative spacer, a para-nitrophenyl (PNP) leaving group, and a maleimide-amido PEG2 spacer for thiol conjugation.
  • Cleavable val-cit dipeptide for enzyme-triggered payload release.
  • PAB self-immolative spacer enables efficient drug release after cleavage.
  • Maleimide-amido PEG2 spacer for thiol-selective conjugation to antibodies.
  • High reported purity suitable for research-grade ADC synthesis.
  • Available in small research sizes for development and testing.
Catalog No. 50-002-03813 Supplier Medchemexpress LLC Supplier No. HY14014750MG
Mal-amido-PEG2-Val-Cit-PAB-PNP is a cleavable two-unit PEG linker designed for use in the synthesis of antibody-drug conjugates (ADCs). It contains a val-cit dipeptide cleavable moiety, a p-aminobenzyl (PAB) self-immolative spacer, a para-nitrophenyl (PNP) leaving group, and a maleimide-amido PEG2 spacer for thiol conjugation.
  • Cleavable val-cit dipeptide for enzyme-triggered payload release.
  • PAB self-immolative spacer enables efficient drug release after cleavage.
  • Maleimide-amido PEG2 spacer for thiol-selective conjugation to antibodies.
  • High reported purity suitable for research-grade ADC synthesis.
  • Available in small research sizes for development and testing.