Learn More
Medchemexpress LLC Mal-amido-PEG2-Val-Cit-PAB-PNP | 2112738-13-1 | MFCD30828687 | 98.4% | 854.86 g·mol⁻1 | C39H50N8O14 | 100 MG
SDP
Catalog No. 5000202220
Encompass_Preferred
Mal-amido-PEG2-Val-Cit-PAB-PNP is a cleavable 2-unit PEG linker used as an intermediate in antibody-drug conjugate (ADC) synthesis. It contains a maleimide thiol-reactive handle, a PEG2 spacer, a valine-citrulline protease-cleavable dipeptide, a para-aminobenzyl self-immolative spacer, and a p-nitrophenyl activated ester for coupling.
  • Cleavable Val-cit peptide for intracellular release
  • Maleimide handle for thiol conjugation
  • PEG2 spacer to improve hydrophilicity and reduce aggregation
  • PNP-activated ester for efficient coupling to nucleophiles
  • Supplied as a solid suitable for research-scale ADC assembly
Catalog No. 50-002-02220 Supplier Medchemexpress LLC Supplier No. HY140147100MG
Mal-amido-PEG2-Val-Cit-PAB-PNP is a cleavable 2-unit PEG linker used as an intermediate in antibody-drug conjugate (ADC) synthesis. It contains a maleimide thiol-reactive handle, a PEG2 spacer, a valine-citrulline protease-cleavable dipeptide, a para-aminobenzyl self-immolative spacer, and a p-nitrophenyl activated ester for coupling.
  • Cleavable Val-cit peptide for intracellular release
  • Maleimide handle for thiol conjugation
  • PEG2 spacer to improve hydrophilicity and reduce aggregation
  • PNP-activated ester for efficient coupling to nucleophiles
  • Supplied as a solid suitable for research-scale ADC assembly