Learn More
Medchemexpress LLC Mal-amido-PEG24-acid | 2171095-70-6 | 98.5% | C58H108N2O29 | 100 MG
SDP
Catalog No. 5000355573
Encompass_Preferred
Mal-amido-PEG24-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules are engineered to contain two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other binds to a specific target protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based linker for PROTAC synthesis
  • Facilitates targeted protein degradation
  • Utilizes the ubiquitin-proteasome system
Catalog No. 50-003-55573 Supplier Medchemexpress LLC Supplier No. HY140975100MG
Mal-amido-PEG24-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules are engineered to contain two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other binds to a specific target protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based linker for PROTAC synthesis
  • Facilitates targeted protein degradation
  • Utilizes the ubiquitin-proteasome system