Learn More
Medchemexpress LLC Mal-amido-PEG24-acid | 2171095-70-6 | 98.5% | C58H108N2O29 | 25 MG
SDP
Catalog No. 5000352901
Encompass_Preferred
Mal-amido-PEG24-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules contain two different ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets a specific protein, leveraging the intracellular ubiquitin-proteasome system for selective protein degradation.
  • Peg-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Intended for research use only
Catalog No. 50-003-52901 Supplier Medchemexpress LLC Supplier No. 25MGHY140975
Mal-amido-PEG24-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules contain two different ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets a specific protein, leveraging the intracellular ubiquitin-proteasome system for selective protein degradation.
  • Peg-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Intended for research use only