Learn More
Medchemexpress LLC Mal-amido-PEG24-acid | 2171095-70-6 | 98.5% | C58H108N2O29 | 50 MG
SDP
Catalog No. 5000353170
Encompass_Preferred
Mal-amido-PEG24-acid is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other for a target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53170 Supplier Medchemexpress LLC Supplier No. HY14097550MG
Mal-amido-PEG24-acid is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other for a target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins