Learn More
Medchemexpress LLC Mal-amido-PEG24-acid | 2171095-70-6 | MFCD13184955 | 98.5% | C58H108N2O29 | 10MG
SDP
Catalog No. 5000261198
Encompass_Preferred
Mal-amido-PEG24-acid is a maleimide-functionalized PEG24 linker with an amido-linked terminal carboxylic acid designed for assembling PROTACs and other bioconjugates. It provides a hydrophilic PEG spacer to improve solubility and a maleimide group for thiol-selective conjugation. CAS 2171095-70-6; molecular weight 1297.47 g·mol⁻1; formula C58H108N2O29.
  • Maleimide group for selective thiol conjugation.
  • Amido-terminated PEG24 spacer with a terminal carboxylic acid for coupling versatility.
  • Hydrophilic spacer improves solubility in aqueous media.
  • High purity (98.5%) suitable for research-grade bioconjugation.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-61198 Supplier Medchemexpress LLC Supplier No. HY14097510MG
Mal-amido-PEG24-acid is a maleimide-functionalized PEG24 linker with an amido-linked terminal carboxylic acid designed for assembling PROTACs and other bioconjugates. It provides a hydrophilic PEG spacer to improve solubility and a maleimide group for thiol-selective conjugation. CAS 2171095-70-6; molecular weight 1297.47 g·mol⁻1; formula C58H108N2O29.
  • Maleimide group for selective thiol conjugation.
  • Amido-terminated PEG24 spacer with a terminal carboxylic acid for coupling versatility.
  • Hydrophilic spacer improves solubility in aqueous media.
  • High purity (98.5%) suitable for research-grade bioconjugation.
  • Stable under recommended storage conditions for long-term use.