Learn More
Medchemexpress LLC Mal-amido-PEG3-acid | 2055353-75-6 | 372.37 | 1G
SDP
Catalog No. 5000350078
Encompass_Preferred
Mal-amido-PEG3-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-50078 Supplier Medchemexpress LLC Supplier No. HY1387271G
Mal-amido-PEG3-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins