Learn More
Medchemexpress LLC Mal-amido-PEG4-acid | 1263045-16-4 | 99.8% | 250 MG
SDP
Catalog No. 5000341452
Encompass_Preferred
Mal-amido-PEG4-acid, also known as 4,7,10,13-Tetraoxa-16-azanonadecanoic acid, 19-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)-17-oxo-, is a high-purity PEG-based PROTAC linker. It is primarily utilized in the synthesis of PROTACs, enabling targeted protein degradation. This compound is characterized by its specific molecular formula C18H28N2O9 and molecular weight of 416.42. It appears as an off-white to yellow solid.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Appears as an off-white to yellow solid
  • Purity of 99.76% by HPLC
Catalog No. 50-003-41452 Supplier Medchemexpress LLC Supplier No. 250MGHY130414
Mal-amido-PEG4-acid, also known as 4,7,10,13-Tetraoxa-16-azanonadecanoic acid, 19-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)-17-oxo-, is a high-purity PEG-based PROTAC linker. It is primarily utilized in the synthesis of PROTACs, enabling targeted protein degradation. This compound is characterized by its specific molecular formula C18H28N2O9 and molecular weight of 416.42. It appears as an off-white to yellow solid.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Appears as an off-white to yellow solid
  • Purity of 99.76% by HPLC