Learn More
Medchemexpress LLC Mal-amido-PEG4-NHS ester | 756525-99-2 | 99.2% | 513.50 | C22H31N3O11 | 100 MG
SDP
Catalog No. 5000301194
Encompass_Preferred
Mal-amido-PEG4-NHS ester is a PEG-based bifunctional linker containing a maleimide handle and an N-hydroxysuccinimide (NHS) ester. It provides a PEG4 spacer for flexibility and solubility and enables orthogonal conjugation to thiols and primary amines for use in PROTAC synthesis and bioconjugation applications.
  • Provides a PEG4 spacer for increased solubility and flexible linker length.
  • Maleimide group enables selective reaction with thiol groups for cysteine conjugation.
  • NHS ester allows rapid coupling to primary amines under mild conditions.
  • High purity suitable for synthetic and medicinal chemistry applications.
  • Stable when stored at -20°C in solid form; follow solvent storage recommendations.
Catalog No. 50-003-01194 Supplier Medchemexpress LLC Supplier No. HYW040134100MG
Mal-amido-PEG4-NHS ester is a PEG-based bifunctional linker containing a maleimide handle and an N-hydroxysuccinimide (NHS) ester. It provides a PEG4 spacer for flexibility and solubility and enables orthogonal conjugation to thiols and primary amines for use in PROTAC synthesis and bioconjugation applications.
  • Provides a PEG4 spacer for increased solubility and flexible linker length.
  • Maleimide group enables selective reaction with thiol groups for cysteine conjugation.
  • NHS ester allows rapid coupling to primary amines under mild conditions.
  • High purity suitable for synthetic and medicinal chemistry applications.
  • Stable when stored at -20°C in solid form; follow solvent storage recommendations.