Learn More
Medchemexpress LLC Mal-amido-PEG4-NHS ester | 756525-99-2 | MFCD11041113 | 99.2% | 513.50 | C22H31N3O11 | 1 G
SDP
Catalog No. 5000299526
Encompass_Preferred
Mal-amido-PEG4-NHS ester is a PEG-based bifunctional linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It is designed for bioconjugation and PROTAC synthesis to connect thiol- and amine-containing ligands while providing a flexible, hydrophilic spacer that can improve solubility and linker reach.
  • Bifunctional: maleimide for thiol coupling and NHS ester for amine coupling.
  • PEG4 spacer increases hydrophilicity and solubility in aqueous media.
  • High purity suitable for research and synthetic applications.
  • Available in multiple pack sizes to support small- and large-scale work.
  • Soluble in DMSO at high concentrations; ultrasonic treatment may aid dissolution.
  • Store frozen for long-term stability; follow solvent storage guidance for solutions.
Catalog No. 50-002-99526 Supplier Medchemexpress LLC Supplier No. HYW0401341G
Mal-amido-PEG4-NHS ester is a PEG-based bifunctional linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It is designed for bioconjugation and PROTAC synthesis to connect thiol- and amine-containing ligands while providing a flexible, hydrophilic spacer that can improve solubility and linker reach.
  • Bifunctional: maleimide for thiol coupling and NHS ester for amine coupling.
  • PEG4 spacer increases hydrophilicity and solubility in aqueous media.
  • High purity suitable for research and synthetic applications.
  • Available in multiple pack sizes to support small- and large-scale work.
  • Soluble in DMSO at high concentrations; ultrasonic treatment may aid dissolution.
  • Store frozen for long-term stability; follow solvent storage guidance for solutions.