Learn More
Medchemexpress LLC HY-140978 100mg Medchemexpress, Mal-amido-PEG4-TFP ester CAS:1807540-84-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY140978100MG
Encompass_Preferred
Medchemexpress, HY-140978 100mg Mal-amido-PEG4-TFP ester CAS:1807540-84-6 Mal-amido-PEG4-TFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1556