Learn More
Medchemexpress LLC Mal-amido-PEG4-TFP ester | 1807540-84-6 | 99.6% | C24H28F4N2O9 | 50 MG
SDP
Catalog No. 5000353173
Encompass_Preferred
Mal-amido-PEG4-TFP ester is a PEG-based PROTAC linker for use in the synthesis of PROTACs. It is intended for research use only. This viscous liquid appears colorless to light yellow and has a molecular weight of 564.48.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Aids in selective degradation of target proteins
  • Viscous liquid appearance
Catalog No. 50-003-53173 Supplier Medchemexpress LLC Supplier No. HY14097850MG
Mal-amido-PEG4-TFP ester is a PEG-based PROTAC linker for use in the synthesis of PROTACs. It is intended for research use only. This viscous liquid appears colorless to light yellow and has a molecular weight of 564.48.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Aids in selective degradation of target proteins
  • Viscous liquid appearance