Learn More
Medchemexpress LLC Mal-amido-PEG5-C2-NHS ester | 1315355-92-0 | >98.0% | 557.55 g·mol⁻1 | C24H35N3O12 | 250 MG
SDP
Catalog No. 5000202450
Encompass_Preferred
Mal-amido-PEG5-C2-NHS ester is a non-cleavable antibody-drug conjugate (ADC) linker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester. The NHS ester labels primary amines on proteins and amine-modified oligonucleotides, while the maleimide provides a thiol-reactive handle; the PEG5 spacer improves solubility and spacing for bioconjugation.
  • Non-cleavable linker with maleimide and NHS reactive groups.
  • PEG5 spacer increases solubility and reduces steric hindrance.
  • NHS ester reacts with primary amines for labeling.
  • Maleimide enables thiol-selective conjugation.
  • High purity suitable for research bioconjugation (≥98.0%).
Catalog No. 50-002-02450 Supplier Medchemexpress LLC Supplier No. HY126508250MG
Mal-amido-PEG5-C2-NHS ester is a non-cleavable antibody-drug conjugate (ADC) linker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester. The NHS ester labels primary amines on proteins and amine-modified oligonucleotides, while the maleimide provides a thiol-reactive handle; the PEG5 spacer improves solubility and spacing for bioconjugation.
  • Non-cleavable linker with maleimide and NHS reactive groups.
  • PEG5 spacer increases solubility and reduces steric hindrance.
  • NHS ester reacts with primary amines for labeling.
  • Maleimide enables thiol-selective conjugation.
  • High purity suitable for research bioconjugation (≥98.0%).