Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-amido-PEG5-C2-NHS ester | 1315355-92-0 | >=98.0% | 557.55 g/mol | C24H35N3O12 | 1 G
SDP

Catalog No. 5000202210
Encompass_Preferred

Mal-amido-PEG5-C2-NHS ester is a non-cleavable PEG-based linker containing a maleimide and an N-hydroxysuccinimide (NHS) ester for conjugation to thiol and amine groups in proteins, peptides, and oligonucleotides. It is commonly used in antibody-drug conjugate (ADC) synthesis and other bioconjugation research applications.

  • Contains maleimide and NHS ester functional groups
  • Non-cleavable PEG-based linker for bioconjugation
  • CAS number 1315355-92-0
  • Molecular weight 557.55 g/mol
  • Formula C24H35N3O12
  • Appearance viscous liquid, colorless to light yellow
  • Purity >=98.0%
  • Storage pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month)
  • Available in 1 G package

Catalog No. 50-002-02210 Supplier Medchemexpress LLC Supplier No. HY1265081G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-amido-PEG5-C2-NHS ester is a non-cleavable PEG-based linker containing a maleimide and an N-hydroxysuccinimide (NHS) ester for conjugation to thiol and amine groups in proteins, peptides, and oligonucleotides. It is commonly used in antibody-drug conjugate (ADC) synthesis and other bioconjugation research applications.

  • Contains maleimide and NHS ester functional groups
  • Non-cleavable PEG-based linker for bioconjugation
  • CAS number 1315355-92-0
  • Molecular weight 557.55 g/mol
  • Formula C24H35N3O12
  • Appearance viscous liquid, colorless to light yellow
  • Purity >=98.0%
  • Storage pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month)
  • Available in 1 G package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.