Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-amido-PEG5-C2-NHS ester | 1315355-92-0 | ≥98.0% | 557.55 g/mol | C24H35N3O12 | 5 G
SDP

Catalog No. 5000202773
Encompass_Preferred

Mal-amido-PEG5-C2-NHS ester is a heterobifunctional PEG5 linker that contains a maleimide group and an N-hydroxysuccinimide (NHS) ester for selective thiol-to-amine conjugation in bioconjugation and linker synthesis.

  • Contains maleimide for thiol conjugation.
  • Contains NHS ester for amine coupling.
  • PEG5 spacer increases hydrophilicity and solubility.
  • Molecular weight approximately 557.55 g/mol.
  • High purity (≥98.0%) suitable for research applications.
  • Available in multiple package sizes, including 5 g.

Catalog No. 50-002-02773 Supplier Medchemexpress LLC Supplier No. HY1265085G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-amido-PEG5-C2-NHS ester is a heterobifunctional PEG5 linker that contains a maleimide group and an N-hydroxysuccinimide (NHS) ester for selective thiol-to-amine conjugation in bioconjugation and linker synthesis.

  • Contains maleimide for thiol conjugation.
  • Contains NHS ester for amine coupling.
  • PEG5 spacer increases hydrophilicity and solubility.
  • Molecular weight approximately 557.55 g/mol.
  • High purity (≥98.0%) suitable for research applications.
  • Available in multiple package sizes, including 5 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.