Learn More
Medchemexpress LLC Mal-amido-PEG6-acid | 1334177-79-5 | 99.3% | 250 MG
SDP
Catalog No. 5000342088
Encompass_Preferred
Mal-amido-PEG6-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. These PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Contains two distinct ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • Other ligand targets the protein of interest
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-42088 Supplier Medchemexpress LLC Supplier No. 250MGHY130399
Mal-amido-PEG6-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. These PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Contains two distinct ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • Other ligand targets the protein of interest
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins