Learn More
Medchemexpress LLC Mal-amido-PEG6-acid | 1334177-79-5 | 99.3% | 50 MG
SDP
Catalog No. 5000338617
Encompass_Preferred
Mal-amido-PEG6-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules are designed to exploit the intracellular ubiquitin-proteasome system, selectively degrading target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Enables targeted protein degradation
  • Facilitates connection of E3 ubiquitin ligase and target protein ligands
Catalog No. 50-003-38617 Supplier Medchemexpress LLC Supplier No. HY13039950MG
Mal-amido-PEG6-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules are designed to exploit the intracellular ubiquitin-proteasome system, selectively degrading target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Enables targeted protein degradation
  • Facilitates connection of E3 ubiquitin ligase and target protein ligands