Learn More
Medchemexpress LLC Mal-amido-PEG6-NHS ester | 1137109-21-7 | 98.76% | 601.60 | 1 G
SDP
Catalog No. 5000452199
Encompass_Preferred
Mal-amido-PEG6-NHS ester is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs themselves consist of two different ligands linked together; one ligand targets an E3 ubiquitin ligase, and the other targets the protein intended for degradation. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
Catalog No. 50-004-52199 Supplier Medchemexpress LLC Supplier No. HYW0088791G
Mal-amido-PEG6-NHS ester is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs themselves consist of two different ligands linked together; one ligand targets an E3 ubiquitin ligase, and the other targets the protein intended for degradation. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs