Learn More
Medchemexpress LLC Mal-amido-PEG6-NHS ester | 1137109-21-7 | 98.76% | 601.60 | 5 G
SDP
Catalog No. 5000453674
Encompass_Preferred
Mal-amido-PEG6-NHS ester is a PEG-based PROTAC linker. It is for research use only. This chemical can be used in the synthesis of PROTACs, which contain two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-53674 Supplier Medchemexpress LLC Supplier No. HYW0088795G
Mal-amido-PEG6-NHS ester is a PEG-based PROTAC linker. It is for research use only. This chemical can be used in the synthesis of PROTACs, which contain two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins