Learn More
Medchemexpress LLC HY-101159 100mg Medchemexpress, Mal-amido-PEG8-acid CAS:1334177-86-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101159100MG
Encompass_Preferred
Medchemexpress, HY-101159 100mg Mal-amido-PEG8-acid CAS:1334177-86-4 Mal-amido-PEG8-acid (example 143) is an ADC linker, extracted from patent US2018339985 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2055