Learn More
Medchemexpress LLC Mal-amido-PEG8-NHS ester | 756525-93-6 | MFCD11041144 | 99.2% | 689.71 g/mol | C30H47N3O15 | 50 MG
SDP
Catalog No. 5000301544
Encompass_Preferred
Mal-amido-PEG8-NHS ester is a bifunctional PEG-based linker bearing a maleimide and an NHS ester. It is designed for bioconjugation workflows to connect thiol- and amine-containing molecules, enabling assembly of PROTACs, antibody drug conjugates, and other conjugates. The PEG8 spacer improves solubility and flexibility, while the reactive termini support efficient, site-selective coupling.
Catalog No. 50-003-01544 Supplier Medchemexpress LLC Supplier No. HYW04013350MG
Mal-amido-PEG8-NHS ester is a bifunctional PEG-based linker bearing a maleimide and an NHS ester. It is designed for bioconjugation workflows to connect thiol- and amine-containing molecules, enabling assembly of PROTACs, antibody drug conjugates, and other conjugates. The PEG8 spacer improves solubility and flexibility, while the reactive termini support efficient, site-selective coupling.