Learn More
Medchemexpress LLC Mal-amido-PEG8-TFP ester | 1924596-31-5 | >98% | C32H44F4N2O13 | 1 G
SDP
Catalog No. 5000355029
Encompass_Preferred
Mal-amido-PEG8-TFP ester is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which facilitate the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-55029 Supplier Medchemexpress LLC Supplier No. HY1409791G
Mal-amido-PEG8-TFP ester is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which facilitate the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only