Learn More
Medchemexpress LLC Mal-amido-PEG8-TFP ester | 1924596-31-5 | C32H44F4N2O13 | 25 MG
SDP
Catalog No. 5000357376
Encompass_Preferred
Mal-amido-PEG8-TFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. This product leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57376 Supplier Medchemexpress LLC Supplier No. 25MGHY140979
Mal-amido-PEG8-TFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. This product leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins