Learn More
Medchemexpress LLC Mal-amido-PEG8-TFP ester | 1924596-31-5 | C32H44F4N2O13 | 250 MG
SDP
Catalog No. 5000357256
Encompass_Preferred
Mal-amido-PEG8-TFP ester is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. PROTACs achieve this by containing two different ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
Catalog No. 50-003-57256 Supplier Medchemexpress LLC Supplier No. 250MGHY140979
Mal-amido-PEG8-TFP ester is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. PROTACs achieve this by containing two different ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins