Learn More
Medchemexpress LLC Mal-amido-PEG9-acid | 2112731-43-6 | 99.54% | 100 MG
SDP
Catalog No. 5000338982
Encompass_Preferred
Mal-amido-PEG9-acid is a high-purity, PEG-based PROTAC linker. It is primarily used in the synthesis of PROTACs, which are compounds designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Purity of 99.54%
Catalog No. 50-003-38982 Supplier Medchemexpress LLC Supplier No. HY130856100MG
Mal-amido-PEG9-acid is a high-purity, PEG-based PROTAC linker. It is primarily used in the synthesis of PROTACs, which are compounds designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Purity of 99.54%