Learn More
Medchemexpress LLC Mal-amido-PEG9-amine (TFA) | 2569576-12-9 | 99.4% | C27H49N3O12.C2HF3O2 | 10MG
SDP
Catalog No. 5000259356
Encompass_Preferred
Mal-amido-PEG9-amine (TFA) is a PEG-based PROTAC linker that contains a maleimide moiety and a terminal amine present as the trifluoroacetic acid salt. It is designed as a hydrophilic spacer to connect ligands in bifunctional molecules and to enable selective thiol conjugation while providing an amine handle for further coupling reactions.
  • Maleimide group enables selective thiol conjugation.
  • Terminal amine (TFA salt) provides an amide-coupling handle.
  • PEG9 spacer increases solubility and molecular flexibility.
  • Suitable for assembly of bifunctional molecules such as PROTACs and bioconjugates.
  • Available in small mg-scale quantities for research use.
Catalog No. 50-002-59356 Supplier Medchemexpress LLC Supplier No. HY140976A10MG
Mal-amido-PEG9-amine (TFA) is a PEG-based PROTAC linker that contains a maleimide moiety and a terminal amine present as the trifluoroacetic acid salt. It is designed as a hydrophilic spacer to connect ligands in bifunctional molecules and to enable selective thiol conjugation while providing an amine handle for further coupling reactions.
  • Maleimide group enables selective thiol conjugation.
  • Terminal amine (TFA salt) provides an amide-coupling handle.
  • PEG9 spacer increases solubility and molecular flexibility.
  • Suitable for assembly of bifunctional molecules such as PROTACs and bioconjugates.
  • Available in small mg-scale quantities for research use.