Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-Bal-Ph(β-D-Glucose)-7-MAD-MDCPT | 2396667-62-0 | 98.2% | 984.87 | C46H44N6O19 | 1 MG
SDP

Catalog No. 5000202816
Encompass_Preferred

Mal-Bal-Ph(β-D-Glucose)-7-MAD-MDCPT is an antibody-drug conjugate (ADC) linker intended for use in the synthesis of ADCs. It is supplied as a light yellow to yellow solid in small quantities for conjugation chemistry and linker development; intended for research use only.

  • ADC linker for synthesizing antibody-drug conjugates.
  • Light yellow to yellow solid appearance.
  • Molecular weight 984.87 and formula C46H44N6O19.
  • High purity (HPLC) 98.19%.
  • Available in small packaging sizes including 1 mg, 5 mg, 10 mg, 50 mg, and 100 mg.
  • Storage: sealed at -20°C for long-term; in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical applications.

Catalog No. 50-002-02816 Supplier Medchemexpress LLC Supplier No. HY1643671MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-Bal-Ph(β-D-Glucose)-7-MAD-MDCPT is an antibody-drug conjugate (ADC) linker intended for use in the synthesis of ADCs. It is supplied as a light yellow to yellow solid in small quantities for conjugation chemistry and linker development; intended for research use only.

  • ADC linker for synthesizing antibody-drug conjugates.
  • Light yellow to yellow solid appearance.
  • Molecular weight 984.87 and formula C46H44N6O19.
  • High purity (HPLC) 98.19%.
  • Available in small packaging sizes including 1 mg, 5 mg, 10 mg, 50 mg, and 100 mg.
  • Storage: sealed at -20°C for long-term; in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.