Learn More
Medchemexpress LLC Mal-Bal-Ph(β-d-glucose)-7-MAD-MDCPT | 2396667-62-0 | 98.2% | 984.87 g/mol | C46H44N6O19 | 10 MG
SDP
Catalog No. 5000202815
Encompass_Preferred
Mal-Bal-Ph(β-D-Glucose)-7-MAD-MDCPT is an antibody-drug conjugate (ADC) linker used in research to attach glucose-derived payloads to antibodies via a maleimide-based handle, enabling targeted bioconjugation and ADC synthesis.
  • Suitable for antibody-drug conjugate synthesis.
  • High purity for research applications.
  • Available in small milligram quantities for method development.
  • Molecular weight 984.87 g/mol and chemical formula C46H44N6O19.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-02815 Supplier Medchemexpress LLC Supplier No. HY16436710MG
Mal-Bal-Ph(β-D-Glucose)-7-MAD-MDCPT is an antibody-drug conjugate (ADC) linker used in research to attach glucose-derived payloads to antibodies via a maleimide-based handle, enabling targeted bioconjugation and ADC synthesis.
  • Suitable for antibody-drug conjugate synthesis.
  • High purity for research applications.
  • Available in small milligram quantities for method development.
  • Molecular weight 984.87 g/mol and chemical formula C46H44N6O19.
  • Intended for research use only; not for human therapeutic use.