Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-bal-ph(β-d-glucose)-7-mad-mdcpt | 2396667-62-0 | 98.2% | 984.87 g/mol | C46H44N6O19 | 5 MG
SDP

Catalog No. 5000202536
Encompass_Preferred

Mal-Bal-Ph(β-D-Glucose)-7-MAD-MDCPT is an ADC linker supplied as a research reagent for use in the synthesis of antibody-drug conjugates and related conjugation chemistry.

  • Can be used to synthesize antibody-drug conjugates (ADCs).
  • Provided in small milligram quantities suitable for research use.
  • Purity reported as 98.19%.
  • Molecular weight 984.87 g/mol and molecular formula C46H44N6O19.
  • Documentation includes a safety data sheet (SDS).

Catalog No. 50-002-02536 Supplier Medchemexpress LLC Supplier No. HY1643675MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mal-Bal-Ph(β-D-Glucose)-7-MAD-MDCPT is an ADC linker supplied as a research reagent for use in the synthesis of antibody-drug conjugates and related conjugation chemistry.

  • Can be used to synthesize antibody-drug conjugates (ADCs).
  • Provided in small milligram quantities suitable for research use.
  • Purity reported as 98.19%.
  • Molecular weight 984.87 g/mol and molecular formula C46H44N6O19.
  • Documentation includes a safety data sheet (SDS).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.