Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-C2-NHS ester | 103750-03-4 | 99.3% | 294.26 g/mol | C13H14N2O6 | 100 MG
SDP

Catalog No. 5000203741
Encompass_Preferred

Mal-C2-NHS ester is a noncleavable antibody-drug conjugate (ADC) linker that contains a maleimide group and an N-hydroxysuccinimide (NHS) ester; it is used to conjugate payloads to antibodies via thiol and amine chemistry during ADC synthesis.

  • Noncleavable ADC linker suitable for antibody conjugation.
  • Contains maleimide and NHS ester reactive groups for thiol and amine coupling.
  • High purity, suitable for research applications.
  • Molecular weight 294.26 g/mol and formula C13H14N2O6.
  • Available in small-scale packages for synthesis and optimization studies.

Catalog No. 50-002-03741 Supplier Medchemexpress LLC Supplier No. HY126502100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-C2-NHS ester is a noncleavable antibody-drug conjugate (ADC) linker that contains a maleimide group and an N-hydroxysuccinimide (NHS) ester; it is used to conjugate payloads to antibodies via thiol and amine chemistry during ADC synthesis.

  • Noncleavable ADC linker suitable for antibody conjugation.
  • Contains maleimide and NHS ester reactive groups for thiol and amine coupling.
  • High purity, suitable for research applications.
  • Molecular weight 294.26 g/mol and formula C13H14N2O6.
  • Available in small-scale packages for synthesis and optimization studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.