Learn More
Medchemexpress LLC Mal-C5-N-bis(PEG2-C2-acid) | 3026691-12-0 | 98.0% | 530.57 | C24H38N2O11 | 100mg
SDP
Catalog No. 5000270289
Encompass_Preferred
Mal-C5-N-bis(PEG2-C2-acid) is an ADC linker used in the synthesis of antibody-drug conjugates (ADCs)[1]
Catalog No. 50-002-70289 Supplier Medchemexpress LLC Supplier No. HYW877850100MG
Mal-C5-N-bis(PEG2-C2-acid) is an ADC linker used in the synthesis of antibody-drug conjugates (ADCs)[1]