Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-C6-α-amanitin | 1578249-76-9 | 98.5% | 1211.34 | C55H78N12O17S | 500 UG
SDP

Catalog No. 5000203163
Encompass_Preferred

Mal-C6-α-Amanitin is an agent-linker conjugate for antibody-drug conjugate (ADC) research that employs α-amanitin (an RNA polymerase II inhibitor) linked via a Mal-C6 maleimide linker to facilitate payload delivery and conjugation studies.

  • Agent-linker conjugate for ADC research
  • Contains α-amanitin, an RNA polymerase II inhibitor
  • Molecular formula C55H78N12O17S, molecular weight 1211.34
  • High purity (≈98.5%)
  • Physical form: white to light yellow solid
  • Storage: protect from light; in solvent: -80°C (6 months), -20°C (1 month)

Catalog No. 50-002-03163 Supplier Medchemexpress LLC Supplier No. HY126683500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-C6-α-Amanitin is an agent-linker conjugate for antibody-drug conjugate (ADC) research that employs α-amanitin (an RNA polymerase II inhibitor) linked via a Mal-C6 maleimide linker to facilitate payload delivery and conjugation studies.

  • Agent-linker conjugate for ADC research
  • Contains α-amanitin, an RNA polymerase II inhibitor
  • Molecular formula C55H78N12O17S, molecular weight 1211.34
  • High purity (≈98.5%)
  • Physical form: white to light yellow solid
  • Storage: protect from light; in solvent: -80°C (6 months), -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.