Learn More
Medchemexpress LLC Mal-C6-amine TFA | 731862-92-3 | MFCD11519191 | 99.7% | 310.27 g/mol | C12H17F3N2O4 | 250 MG
SDP
Catalog No. 5000302588
Encompass_Preferred
Mal-C6-amine (TFA) is an alkyl chain-based PROTAC linker supplied as the trifluoroacetate salt for use in the synthesis of proteolysis-targeting chimeras. It provides a maleimide-reactive terminus and a six-carbon spacer that enable thiol conjugation and optimized linker length for degrader constructs.
  • Maleimide-reactive terminus for selective conjugation to thiols.
  • C6 alkyl spacer provides flexibility and appropriate linker length.
  • Trifluoroacetate salt form for improved handling and solubility.
  • High purity suitable for chemical synthesis (listed as 99.73%).
  • Soluble in DMSO for convenient formulation and reactions.
Catalog No. 50-003-02588 Supplier Medchemexpress LLC Supplier No. 250MGHYW018291
Mal-C6-amine (TFA) is an alkyl chain-based PROTAC linker supplied as the trifluoroacetate salt for use in the synthesis of proteolysis-targeting chimeras. It provides a maleimide-reactive terminus and a six-carbon spacer that enable thiol conjugation and optimized linker length for degrader constructs.
  • Maleimide-reactive terminus for selective conjugation to thiols.
  • C6 alkyl spacer provides flexibility and appropriate linker length.
  • Trifluoroacetate salt form for improved handling and solubility.
  • High purity suitable for chemical synthesis (listed as 99.73%).
  • Soluble in DMSO for convenient formulation and reactions.