Learn More
Medchemexpress LLC Mal-C6-amine (TFA) | 731862-92-3 | MFCD11519191 | 99.7% | 310.27 g/mol | C12H17F3N2O4 | 5 G
SDP
Catalog No. 5000299900
Encompass_Preferred
Mal-C6-amine (TFA) is an alkyl chain-based PROTAC linker (maleimide C6 amine, trifluoroacetate salt) supplied for research use. It facilitates thiol conjugation via the maleimide group and is provided as an off-white to light yellow solid.
  • Alkyl chain-based linker for PROTAC synthesis.
  • Maleimide functional group enables thiol conjugation.
  • Trifluoroacetate salt form for improved stability and handling.
  • High purity (~99.7%) suitable for research applications.
  • Soluble in DMSO (100 mg/mL) with ultrasonic assistance.
  • Store sealed at 4°C; in solution keep at -80°C for long-term storage.
  • For research use only; not for human use.
Catalog No. 50-002-99900 Supplier Medchemexpress LLC Supplier No. HYW0182915G
Mal-C6-amine (TFA) is an alkyl chain-based PROTAC linker (maleimide C6 amine, trifluoroacetate salt) supplied for research use. It facilitates thiol conjugation via the maleimide group and is provided as an off-white to light yellow solid.
  • Alkyl chain-based linker for PROTAC synthesis.
  • Maleimide functional group enables thiol conjugation.
  • Trifluoroacetate salt form for improved stability and handling.
  • High purity (~99.7%) suitable for research applications.
  • Soluble in DMSO (100 mg/mL) with ultrasonic assistance.
  • Store sealed at 4°C; in solution keep at -80°C for long-term storage.
  • For research use only; not for human use.