Learn More
Medchemexpress LLC HY-128711 5mg Medchemexpress, MAL-di-EG-Val-Cit-PAB-MMAF CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1287115MG
Encompass_Preferred
Medchemexpress, HY-128711 5mg MAL-di-EG-Val-Cit-PAB-MMAF CAS: MAL-di-EG-Val-Cit-PAB-MMAF consists the ADCs linker (MAL-di-EG-Val-Cit-PAB) and potent tubulin polymerization blocker (MMAF, Monomethyl auristatin F) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2631