Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-di-EG-Val-Cit-PAB-MMAF | 98.6% | 1476.75 g·mol⁻¹ | C73H113N13O19 | 10 MG
SDP

Catalog No. 5000203264
Encompass_Preferred

MAL-di-EG-Val-Cit-PAB-MMAF is an antibody-drug conjugate (ADC) drug-linker conjugate that couples a cathepsin-cleavable Val-Cit-PAB linker to monomethyl auristatin F (MMAF), a potent tubulin polymerization inhibitor. It is supplied as a solid for use in ADC synthesis, conjugation studies, and preclinical research.

  • Molecular formula: C73H113N13O19 (manufacturer SDS; alternate product page reports a different formula)
  • Molecular weight: 1476.75 g·mol⁻¹ (manufacturer SDS; product page lists a different value)
  • Purity: manufacturer product page reports ~98.6% (alternate data-sheet entry lists ~96.0%); verify batch certificate for exact lot purity
  • Physical form: solid, unstable in solution; prepare fresh for use and store under recommended conditions
  • Typical use: building block for antibody-drug conjugate synthesis and preclinical ADC research
  • Available pack sizes: includes 10 MG; larger sizes available by quote

Catalog No. 50-002-03264 Supplier Medchemexpress LLC Supplier No. HY12871110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MAL-di-EG-Val-Cit-PAB-MMAF is an antibody-drug conjugate (ADC) drug-linker conjugate that couples a cathepsin-cleavable Val-Cit-PAB linker to monomethyl auristatin F (MMAF), a potent tubulin polymerization inhibitor. It is supplied as a solid for use in ADC synthesis, conjugation studies, and preclinical research.

  • Molecular formula: C73H113N13O19 (manufacturer SDS; alternate product page reports a different formula)
  • Molecular weight: 1476.75 g·mol⁻¹ (manufacturer SDS; product page lists a different value)
  • Purity: manufacturer product page reports ~98.6% (alternate data-sheet entry lists ~96.0%); verify batch certificate for exact lot purity
  • Physical form: solid, unstable in solution; prepare fresh for use and store under recommended conditions
  • Typical use: building block for antibody-drug conjugate synthesis and preclinical ADC research
  • Available pack sizes: includes 10 MG; larger sizes available by quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.