Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-GGG-Bal-NHS ester | 1193111-65-7 | 97.6% | 522.47 g/mol | 5 MG
SDP

Catalog No. 5000201930
Encompass_Preferred

Mal-GGG-Bal-NHS ester is an ADC linker reagent used to introduce a maleimide-containing GGG-Bal-NHS moiety for conjugation to antibodies or other amine- or thiol-containing biomolecules.

  • Used as an ADC linker in antibody-drug conjugate synthesis.
  • Contains an NHS ester for amine coupling and a maleimide for thiol conjugation.
  • High reported purity of 97.6% for reliable coupling performance.
  • Soluble in water at high concentration (50 mg/mL) with ultrasonic treatment if needed.
  • Recommended storage under inert atmosphere and cold conditions to preserve stability.

Catalog No. 50-002-01930 Supplier Medchemexpress LLC Supplier No. HY1604085MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-GGG-Bal-NHS ester is an ADC linker reagent used to introduce a maleimide-containing GGG-Bal-NHS moiety for conjugation to antibodies or other amine- or thiol-containing biomolecules.

  • Used as an ADC linker in antibody-drug conjugate synthesis.
  • Contains an NHS ester for amine coupling and a maleimide for thiol conjugation.
  • High reported purity of 97.6% for reliable coupling performance.
  • Soluble in water at high concentration (50 mg/mL) with ultrasonic treatment if needed.
  • Recommended storage under inert atmosphere and cold conditions to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.