Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-gly-pab-exatecan-d-glucuronic acid | 2763252-25-9 | 99.8% | 984.89 g/mol | C47H45FN6O17 | 50 MG
SDP

Catalog No. 5000204326
Encompass_Preferred

Mal-Gly-PAB-Exatecan-D-glucuronic acid is an ADC intermediate used in antibody-drug conjugate synthesis and tumor research. It is supplied as a light yellow to green-yellow solid with high purity and is formulated for improved solubility via glucuronidation.

  • Purity 99.8%.
  • Molecular weight 984.89 g/mol.
  • Chemical formula C47H45FN6O17.
  • Solubility DMSO 100 mg/mL; ultrasonic assistance recommended.
  • Recommended storage solid at 4°C, protect from light, under nitrogen.
  • Intended use for research and ADC synthesis; not for clinical use.

Catalog No. 50-002-04326 Supplier Medchemexpress LLC Supplier No. HY15317950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-Gly-PAB-Exatecan-D-glucuronic acid is an ADC intermediate used in antibody-drug conjugate synthesis and tumor research. It is supplied as a light yellow to green-yellow solid with high purity and is formulated for improved solubility via glucuronidation.

  • Purity 99.8%.
  • Molecular weight 984.89 g/mol.
  • Chemical formula C47H45FN6O17.
  • Solubility DMSO 100 mg/mL; ultrasonic assistance recommended.
  • Recommended storage solid at 4°C, protect from light, under nitrogen.
  • Intended use for research and ADC synthesis; not for clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.