Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-Gly-PAB-Exatecan-D-glucuronic acid | 2763252-25-9 | 99.8% | 984.9 g/mol | C47H45FN6O17 | 10 MG
SDP

Catalog No. 5000204487
Encompass_Preferred

Mal-Gly-PAB-Exatecan-D-glucuronic acid is a chemical intermediate designed for use in antibody-drug conjugate (ADC) synthesis and related tumor research. It features a maleimide-linked Gly-PAB exatecan payload coupled to a D-glucuronic acid moiety to enable enzymatically cleavable linker strategies. Supplied as a high-purity solid for laboratory research use only.

  • Suitable for ADC synthesis and drug-linker development.
  • Contains a D-glucuronic acid enzymatically cleavable linker.
  • High purity suitable for research applications (99.8%).
  • Available in small-scale and bulk quantities depending on project needs.
  • Solid, light yellow to green yellow appearance for easy handling.
  • Molecular weight ~984.9 g/mol; formula C47H45FN6O17.

Catalog No. 50-002-04487 Supplier Medchemexpress LLC Supplier No. HY15317910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-Gly-PAB-Exatecan-D-glucuronic acid is a chemical intermediate designed for use in antibody-drug conjugate (ADC) synthesis and related tumor research. It features a maleimide-linked Gly-PAB exatecan payload coupled to a D-glucuronic acid moiety to enable enzymatically cleavable linker strategies. Supplied as a high-purity solid for laboratory research use only.

  • Suitable for ADC synthesis and drug-linker development.
  • Contains a D-glucuronic acid enzymatically cleavable linker.
  • High purity suitable for research applications (99.8%).
  • Available in small-scale and bulk quantities depending on project needs.
  • Solid, light yellow to green yellow appearance for easy handling.
  • Molecular weight ~984.9 g/mol; formula C47H45FN6O17.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.