Learn More
MSC Malish Mal-Grit Lite Brush, 14 in, Nylon/SiC, Fine, 1 ea
Catalog No. 502819200
Encompass
The Mal-Grit Lite Rotary Brush features self-replenishing silicone carbide impregnated nylon bristles, offering a lifespan that outlasts floor pads over 100 to 1. Designed for wet environments, it provides the durability of grit brushes with minimum aggression for daily scrubbing of various surfaces like tiles, marble, terrazzo, and non-slip epoxy finishes.
  • Water and chemical resistant TUFF-BLOCK for maximum life expectancy
  • Trim length of 1.5 in.
  • Includes NP-47 clutch plate
  • Brush diameter of 14 in.
  • Fine grit grade
Catalog No. 50-281-9200 Supplier MSC Supplier No. 15839814
The Mal-Grit Lite Rotary Brush features self-replenishing silicone carbide impregnated nylon bristles, offering a lifespan that outlasts floor pads over 100 to 1. Designed for wet environments, it provides the durability of grit brushes with minimum aggression for daily scrubbing of various surfaces like tiles, marble, terrazzo, and non-slip epoxy finishes.
  • Water and chemical resistant TUFF-BLOCK for maximum life expectancy
  • Trim length of 1.5 in.
  • Includes NP-47 clutch plate
  • Brush diameter of 14 in.
  • Fine grit grade