Learn More
Medchemexpress LLC Mal-L-PA-NH-Boc | 1491152-23-8 | 99.3% | 284.27 g/mol | C12H16N2O6 | 50 MG
SDP
Catalog No. 5000203576
Encompass_Preferred
Mal-L-PA-NH-Boc is a noncleavable antibody-drug conjugate (ADC) linker used to attach cytotoxic payloads to antibodies during ADC synthesis. It is supplied as a white to off-white solid intended for research use.
  • Noncleavable ADC linker for stable payload attachment.
  • High purity (99.3%).
  • White to off-white solid physical form.
  • Storage stability: powder stable at -20°C for 3 years and 4°C for 2 years; in solvent stable at -80°C for 6 months and -20°C for 1 month.
  • Includes CAS number 1491152-23-8 and molecular weight 284.27 g/mol.
Catalog No. 50-002-03576 Supplier Medchemexpress LLC Supplier No. HY4191950MG
Mal-L-PA-NH-Boc is a noncleavable antibody-drug conjugate (ADC) linker used to attach cytotoxic payloads to antibodies during ADC synthesis. It is supplied as a white to off-white solid intended for research use.
  • Noncleavable ADC linker for stable payload attachment.
  • High purity (99.3%).
  • White to off-white solid physical form.
  • Storage stability: powder stable at -20°C for 3 years and 4°C for 2 years; in solvent stable at -80°C for 6 months and -20°C for 1 month.
  • Includes CAS number 1491152-23-8 and molecular weight 284.27 g/mol.