Learn More
Medchemexpress LLC Mal-N-bis(PEG4-C2-acid) | 2100306-52-1 | 98.0% | C29H48N2O15 | 50 MG
SDP
Catalog No. 5000356903
Encompass_Preferred
Mal-N-bis(PEG4-C2-acid) is a PROTAC linker utilized in laboratory chemical applications and the manufacture of substances. This high-purity compound features a molecular formula of C29H48N2O15 and a molecular weight of 664.70. It is suitable for various chemical synthesis projects, offering stability under recommended storage conditions for both pure and dissolved forms.
  • High purity (98.0%)
  • Functions as a PROTAC linker
  • Suitable for laboratory chemical applications
  • Stable for up to 3 years in pure form at -20°C
  • Stable for up to 6 months in solvent at -80°C
Catalog No. 50-003-56903 Supplier Medchemexpress LLC Supplier No. HY14053050MG
Mal-N-bis(PEG4-C2-acid) is a PROTAC linker utilized in laboratory chemical applications and the manufacture of substances. This high-purity compound features a molecular formula of C29H48N2O15 and a molecular weight of 664.70. It is suitable for various chemical synthesis projects, offering stability under recommended storage conditions for both pure and dissolved forms.
  • High purity (98.0%)
  • Functions as a PROTAC linker
  • Suitable for laboratory chemical applications
  • Stable for up to 3 years in pure form at -20°C
  • Stable for up to 6 months in solvent at -80°C