Learn More
Medchemexpress LLC Mal-NH-ethyl-SS-propionic acid | 2128735-24-8 | MFCD31536759 | 99.9% | 332.40 | C12H16N2O5S2 | 100 MG
SDP
Catalog No. 5000202098
Encompass_Preferred
Mal-NH-ethyl-SS-propionic acid is a cleavable antibody-drug conjugate (ADC) linker containing a maleimide reactive group and a disulfide-containing spacer. It is used to couple payloads to antibodies via thiol-maleimide chemistry and enables reductive intracellular release of the payload.
  • Cleavable disulfide linker for reductive intracellular release.
  • Maleimide group for efficient thiol conjugation.
  • High purity solid suitable for research applications.
  • Molecular formula C12H16N2O5S2, molecular weight 332.40.
  • Available in multiple pack sizes, including 100 MG.
Catalog No. 50-002-02098 Supplier Medchemexpress LLC Supplier No. HY140120100MG
Mal-NH-ethyl-SS-propionic acid is a cleavable antibody-drug conjugate (ADC) linker containing a maleimide reactive group and a disulfide-containing spacer. It is used to couple payloads to antibodies via thiol-maleimide chemistry and enables reductive intracellular release of the payload.
  • Cleavable disulfide linker for reductive intracellular release.
  • Maleimide group for efficient thiol conjugation.
  • High purity solid suitable for research applications.
  • Molecular formula C12H16N2O5S2, molecular weight 332.40.
  • Available in multiple pack sizes, including 100 MG.