Learn More
Medchemexpress LLC Mal-NH-ethyl-SS-propionic acid | 2128735-24-8 | MFCD31536759 | 99.9% | 332.40 | C12H16N2O5S2 | 25 MG
SDP
Catalog No. 5000202093
Encompass_Preferred
Mal-NH-ethyl-SS-propionic acid is a cleavable antibody-drug conjugate (ADC) linker designed for thiol conjugation and reductive release of payloads in target cells. It contains a maleimide reactive group, a disulfide cleavable spacer, and a terminal propionic acid, and is supplied as a solid for research use only.
  • Cleavable disulfide spacer enables reductive release of payloads inside cells.
  • Maleimide functional group reacts selectively with thiol groups for conjugation.
  • Terminal carboxylic acid provides a handle for further functionalization.
  • High purity suitable for research applications.
  • Available in multiple small-scale pack sizes for lab use.
Catalog No. 50-002-02093 Supplier Medchemexpress LLC Supplier No. HY14012025MG
Mal-NH-ethyl-SS-propionic acid is a cleavable antibody-drug conjugate (ADC) linker designed for thiol conjugation and reductive release of payloads in target cells. It contains a maleimide reactive group, a disulfide cleavable spacer, and a terminal propionic acid, and is supplied as a solid for research use only.
  • Cleavable disulfide spacer enables reductive release of payloads inside cells.
  • Maleimide functional group reacts selectively with thiol groups for conjugation.
  • Terminal carboxylic acid provides a handle for further functionalization.
  • High purity suitable for research applications.
  • Available in multiple small-scale pack sizes for lab use.