Learn More
Medchemexpress LLC Mal-NH-ethyl-SS-propionic acid | 2128735-24-8 | MFCD31536759 | 99.9% | 332.40 g/mol | C12H16N2O5S2 | 50 MG
SDP
Catalog No. 5000202651
Encompass_Preferred
Mal-NH-ethyl-SS-propionic acid is a cleavable antibody-drug conjugate (ADC) linker containing a maleimide reactive group and a disulfide-containing spacer that enables conjugation to thiol groups and reductive payload release under intracellular reducing conditions.
  • Contains maleimide reactive group for thiol conjugation
  • Disulfide-containing spacer enables cleavable release under reducing conditions
  • Suitable for synthesis of antibody-drug conjugates and linker-payload constructs
  • High purity for research applications
  • Available in small package sizes for lab-scale synthesis
Catalog No. 50-002-02651 Supplier Medchemexpress LLC Supplier No. HY14012050MG
Mal-NH-ethyl-SS-propionic acid is a cleavable antibody-drug conjugate (ADC) linker containing a maleimide reactive group and a disulfide-containing spacer that enables conjugation to thiol groups and reductive payload release under intracellular reducing conditions.
  • Contains maleimide reactive group for thiol conjugation
  • Disulfide-containing spacer enables cleavable release under reducing conditions
  • Suitable for synthesis of antibody-drug conjugates and linker-payload constructs
  • High purity for research applications
  • Available in small package sizes for lab-scale synthesis