Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-NH-PEG8-Boc | 2864817-39-8 | >98.0% | 648.74 | 500 MG
SDP

Catalog No. 5000348631
Encompass_Preferred

Mal-NH-PEG8-Boc is a PEG-based PROTAC linker, a chemical compound with the molecular formula C30H52N2O13 and a molecular weight of 648.74. This linker is designed for use in the synthesis of PROTACs, which are bifunctional molecules that induce the degradation of target proteins. It is generally supplied as an oil with a purity greater than 98%. The product is recommended to be stored at 4°C under nitrogen to maintain its stability.

  • Functions as a PROTAC linker
  • PEG-based structure enhances solubility
  • Chemical formula: C30H52N2O13
  • Molecular weight: 648.74
  • Purity typically greater than 98%
  • Appears as an oil
  • Used in the synthesis of PROTACs

Catalog No. 50-003-48631 Supplier Medchemexpress LLC Supplier No. HY138447500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-NH-PEG8-Boc is a PEG-based PROTAC linker, a chemical compound with the molecular formula C30H52N2O13 and a molecular weight of 648.74. This linker is designed for use in the synthesis of PROTACs, which are bifunctional molecules that induce the degradation of target proteins. It is generally supplied as an oil with a purity greater than 98%. The product is recommended to be stored at 4°C under nitrogen to maintain its stability.

  • Functions as a PROTAC linker
  • PEG-based structure enhances solubility
  • Chemical formula: C30H52N2O13
  • Molecular weight: 648.74
  • Purity typically greater than 98%
  • Appears as an oil
  • Used in the synthesis of PROTACs

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.